missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Mouse Recombinant IL-13

Product Code. 17189391
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
This item is not returnable. View return policy

Product Code. 17189391

Brand: STEMCELL Technologies 78030.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 13

  • Interleukin 13 (IL-13) is a type I cytokine, and signals through type I cytokine receptors to activate JAK/STAT and IRS-1/IRS-2 pathways
  • IL-13 is produced by T cells and innate lymphoid cells (Pulendran & Artis) and it inhibits production of pro-inflammatory cytokines, prostaglandins, and reactive oxygen and nitrogen species by monocytes and macrophages (Hershey)
  • Unlike human B cells, IL-13 has no effect on mouse B cell development and function
  • IL-13 promotes eosinophil survival, activation, and recruitment, and also activates mast cells (Hershey)
  • IL-13 regulates gastrointestinal parasite expulsion, airway hyperresponsiveness, allergic inflammation, tissue eosinophilia, goblet cell hyperplasia, intracellular parasitism, tissue remodeling, tumor cell growth, and fibrosis
  • IL-13 induces hypercontractility of smooth muscles by directly acting on the muscle cells as well as the enteric nerves (Wynn)

Specifications

Name Interleukin-13, NC30
Quantity 10 μg
Cell Type B Cells; Hematopoietic Stem and Progenitor Cells; Monocytes
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.