missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant IL-11
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
Description
- Interleukin 11 (IL-11) is a pleiotropic cytokine with effects on various tissues including the bone marrow, brain, and intestinal mucosa
- It belongs to the IL-6 family of cytokines that share a common signal transducer, gp130
- Culture of mouse bone marrow cells with IL-11 in combination with IL-3, IL-6, and stem cell factor induces significant expansion and proliferation of colony-forming cells in vitro
- In addition, in combination with IL-3, IL-11 significantly enhances the growth of megakaryocytic colonies in vitro, suggesting its role in augmenting mouse megakaryopoiesis
- IL-11 is expressed in a wide range of normal adult mouse tissues, including the central nervous system, thymus, lung, and bone
- The mouse IL-11 cDNA was cloned using an expression library generated from the lipopolysaccharide-induced mouse fetal thymic cell line, T2
Specifications
Specifications
| Name | Adipogenic inhibitory factor, AGIF, Interleukin-11, Oprelvekin |
| Quantity | 100 μg |
| Cell Type | B Cells; Hematopoietic Stem and Progenitor Cells; Other; Pluripotent Stem Cells; T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction