missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Mouse Recombinant IL-11

Product Code. 17149381
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
This item is not returnable. View return policy

Product Code. 17149381

Brand: STEMCELL Technologies 78026

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 11

  • Interleukin 11 (IL-11) is a pleiotropic cytokine with effects on various tissues including the bone marrow, brain, and intestinal mucosa
  • It belongs to the IL-6 family of cytokines that share a common signal transducer, gp130
  • Culture of mouse bone marrow cells with IL-11 in combination with IL-3, IL-6, and stem cell factor induces significant expansion and proliferation of colony-forming cells in vitro
  • In addition, in combination with IL-3, IL-11 significantly enhances the growth of megakaryocytic colonies in vitro, suggesting its role in augmenting mouse megakaryopoiesis
  • IL-11 is expressed in a wide range of normal adult mouse tissues, including the central nervous system, thymus, lung, and bone
  • The mouse IL-11 cDNA was cloned using an expression library generated from the lipopolysaccharide-induced mouse fetal thymic cell line, T2

Specifications

Name Adipogenic inhibitory factor, AGIF, Interleukin-11, Oprelvekin
Quantity 100 μg
Cell Type B Cells; Hematopoietic Stem and Progenitor Cells; Other; Pluripotent Stem Cells; T Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.