missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Mouse Recombinant IFN-gamma

Product Code. p-7217559
Encompass_Preferred
Click to view available options
Quantity:
100 μg
1000 μg
20 μg
Unit Size:
1000µg
100µg
20µg
This item is not returnable. View return policy

Product Code. 15806044

Brand: STEMCELL Technologies 78021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interferon-gamma

  • Interferon-gamma (IFN-γ), also known as type II interferon, is produced by T and NK cells, and in smaller amounts by dendritic cells and macrophages
  • IFN-γ is controlled by cytokines such as IL-12 and IL-18 secreted in response to infection
  • IFN-γ binds to a receptor complex and initiates signal transduction via the JAK/STAT pathway; this culminates in the transcription and activation of many genes that control a diverse array of immunological functions
  • IFN-γ stimulates the antimicrobial and anti-tumor activity of macrophages, NK cells, and neutrophils by promoting the activation of microbial effector functions such as production of reactive oxygen species, NO intermediates, complement, etc
  • IFN-γ enhances MHC class I and II expression in dendritic cells and mononuclear phagocytes, as well as the production of IL-12 by dendritic cells

Specifications

Name Interferon gamma, Type II interferon
Quantity 100 μg
Cell Type B Cells; Hematopoietic Stem and Progenitor Cells; Intestinal Cells; Mesenchymal Stem and Progenitor Cells; T Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.