missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse/Rat WISP-1/CCN4 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (5 Plates)
Unit Size:
1 set
Description
The Mouse/Rat WISP-1/CCN4 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse, rat WISP-1/CCN4.
Specifications
Specifications
| Assay Range | 15.60 - 1000pg/mL |
| Target Species | Mouse, Rat |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 8840 |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction