missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat TSC22 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3779-SP
This item is not returnable.
View return policy
Description
TSC22/TSC22D1 Polyclonal specifically detects TSC22/TSC22D1 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| TSC22/TSC22D1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cerebral protein 2, Egr5, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22D1, TSC22DKFZp686O19206 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8848 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| P62500.2 | |
| TSC22D1 | |
| E. coli-derived recombinant mouse TSC22 Met1-Ala143 Accession # P62500.2 | |
| 25 μg | |
| Primary | |
| Detects mouse and rat TSC22 in direct ELISAs and Western blots. | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction