Learn More
Mouse/Rat Tenascin R Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 3 publications
Brand:  R&D Systems MAB1624-SP
Product Code. 18769986
  • £170.00 / 25µg
Description
Tenascin R Monoclonal specifically detects Tenascin R in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tenascin R
Monoclonal
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Janusin, MGC149328, Restrictin, tenascin R (restrictin, janusin), tenascin-R, TNR, TN-R
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7143
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot, Immunohistochemistry
619
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
TNR
ôrest L2ö fraction from bovine brain
25 μg
Primary
Detects mouse and rat Tenascin R in Western blots.
Mouse, Rat
Purified