missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat RIBP/SH2D2A Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6265
This item is not returnable.
View return policy
Description
SH2D2A Polyclonal specifically detects SH2D2A in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SH2D2A | |
| Polyclonal | |
| Unconjugated | |
| Q9QXK9 | |
| SH2D2A | |
| E. coli-derived recombinant mouse RIBP/SH2D2A Ile299-Ser374 Accession # Q9QXK9 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat RIBP/SH2D2A in Western blots. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| F2771, RIBP, SCAP, SH2 domain containing 2A, SH2 domain protein 2A, SH2 domain-containing adapter protein, T cell specific adapter protein TSAd, T cell-specific adapter protein, T lymphocyte specific adaptor protein, TSAd, TSADSH2 domain-containing protein 2A, VEGF receptor-associated protein, VRAP | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9047 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction