missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat RAGE Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 15 publications
£189.00 - £1860.00
Specifications
| Antigen | RAGE |
|---|---|
| Clone | 175410 |
| Concentration | LYOPH |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18705568
![]() |
R&D Systems
MAB1179-SP |
25 μg |
£189.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18754387
![]() |
R&D Systems
MAB1179-100 |
100 μg |
£480.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18704368
![]() |
R&D Systems
MAB1179-500 |
500 μg |
£1860.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGER Monoclonal specifically detects AGER in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| RAGE | |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
| Ager | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 175410 | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse, Rat | |
| NP_031451 | |
| 5891 | |
| Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Ala342 Accession # NP_031451 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse and rat RAGE in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human RAGE is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title