missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat Phospho-DARPP-32 (T34) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF2847-SP
This item is not returnable.
View return policy
Description
DARPP-32 Polyclonal specifically detects DARPP-32 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| DARPP-32 (p thr34) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PPP1R1B | |
| Phosphopeptide containing mouse DARPP-32 T34 site | |
| 25 μg | |
| Primary | |
| Detects mouse and rat DARPP-32 when phosphorylated at T34 in Western blots. There is weak cross-reactivity with phosphorylated PP1 Inhibitor-1, which contains a nearly identical phosphorylation site. Reactivity with human Phospho-DARPP-32 has not been confirmed. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| DARPP32, PPP1R1B, protein phosphatase 1, regulatory (inhibitor) subunit 1B, regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein, DARPP-32) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84152 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction