missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat Neuropilin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 12 publications
Brand: R&D Systems AF567
This item is not returnable.
View return policy
Description
Neuropilin-2 Polyclonal specifically detects Neuropilin-2 in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Blockade of Receptor-ligand Interaction.
Specifications
| Neuropilin-2 | |
| Polyclonal | |
| Unconjugated | |
| O35276 | |
| NRP2 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-2 Gln23-Asp857 (Val809-Asp825 del) Accession # O35276 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat Neuropilin-2 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human Neuropilin-2 and recombinant rat Neuropilin-1 is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
| MGC126574, neuropilin 2, neuropilin-2, Neuropilin2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8828 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction