missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat M-Cadherin/Cadherin-15 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB76771-SP
This item is not returnable.
View return policy
Description
M-Cadherin/Cadherin-15 Monoclonal specifically detects M-Cadherin/Cadherin-15 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| M-Cadherin/Cadherin-15 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P33146 | |
| CDH15 | |
| Mouse myeloma cell line NS0-derived recombinant mouse M-Cadherin/Cadherin-15 Val22-Ala605 Accession # P33146 | |
| 25 μg | |
| Primary | |
| Detects mouse M-Cadherin/Cadherin-15 in direct ELISAs and mouse and rat M-Cadherin/Cadherin-15 in Western blots. In direct ELISAs, no cross-reactivity with recombinant human M-Cadherin, recombinant mouse (rm) Cadherin-13, or rmP-Cadherin is observed. | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| 800512 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cadherin 15, M-cadherin (myotubule), cadherin 15, type 1, M-cadherin (myotubule), Cadherin-14, Cadherin-15, cadherin-3, CDH14, CDH14cadherin-14, CDH15, CDH3cadherin-15, CDHM, MCAD, M-cadherin, MCadherin, MRD3, Muscle cadherin, muscle-cadherin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1013 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction