missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse/Rat Leptin Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
1 Pk. (6 Plates)
Unit Size:
192 tests
Pack of 96
Specifications
Specifications
| Assay Range | 31.30 to 2000 pg/mL |
| Assay Sensitivity | 5.56 pg/mL |
| Sample Type | Cell Culture Supernatant, Mouse Plasma (EDTA), Mouse Plasma (Heparin), Mouse Serum, Rat Plasma (EDTA), Rat Plasma (Heparin), Rat Serum, Tissue Homogenates |
| Specificity | Natural and Recombinant Mouse and Rat Leptin |
| Target Species | Mouse, Rat |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3952 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction