missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse/Rat IGF1 ELISA Kit

Product Code. 16831626 Shop All Proteintech Products
Click to view available options
Target Species:
Mouse, Rat
missing translation for 'unitSize'
96 tests
Questo articolo non è restituibile. Consulta la politica di reso

Codice prodotto. 16831626

missing translation for 'mfr': Proteintech™ KE1003296T

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

KE10032 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The Mouse/Rat IGF1 ELISA kit is to be used to detect and quantify protein levels of endogenous mouse/rat IGF1.

  • The assay recognizes mouse/rat IGF1. An antibody specific for mouse/rat IGF1 has been pre-coated onto the microwells. The mouse/rat IGF1 protein in samples is captured by the coated antibody after incubation.
  • Following extensive washing, another antibody of biotinylated specific for mouse/rat IGF1 is added to detect the captured mouse/rat IGF1 protein.
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
  • Insulin-like-growth factor 1 (IGF1), a 70 amino-acid peptide hormone is the principal mediator of biochemical effects of growth hormone (GH). IGF1 is an important growth factor in the regulation of cell proliferation and differentiation.
  • IGF1 is largely synthesized in the liver (75%) and, to a lesser extent, in peripheral tissues. IGF1 have been shown to play an essential role in preventing the formation of fatty liver. IGF1 is a potent mitogen and is inhibited by IGF-binding protein-3 (IGFBP3). High serum IGF1 and low IGFBP3 are associated with increased risk of several carcinomas.
  • Mice lacking IGF1 exhibit generalized organ hypoplasia including underdevelopment of the central nervous system and developmental defects in bone, muscle and reproductive systems.

Specifica

Accession Number BC012409
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 1.3 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Mouse, Rat
Gene ID (Entrez) 16000
Host Species Mouse/Rat
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P05017
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • 96TEST Mouse CXCL2 ELISA Kit
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Consigli di prudenza
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informazioni supplementari
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato