missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse/Rat IGF-I/IGF-1 Quantikine ELISA Kit

Product Code. 18742384
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18742384

Brand: R&D Systems™ SMG100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse/Rat IGF-I/IGF-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat IGF-I/IGF-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat IGF-I/IGF-1 in cell culture supernates (25 ul), tissue homogenates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 8.4pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type IGF-I Quantikine ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and Recombinant Mouse and Rat IGF-I
Sufficient For 6 x 96-well plates
Target Species Mouse, Rat
For Use With (Application) ELISA
Gene ID (Entrez) 24482
Quantity 1 Kit
Sample Volume Cell Culture Supernates (25uL),Tissue Homogenates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.