missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse/Rat Flt-3 Ligand/FLT3L ELISA Kit

Product Code. 17751499 Shop All Proteintech Products
Click to view available options
Quantity:
1 Kit
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 17751499

Brand: Proteintech™ KE1007296T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Mouse/Rat Flt-3 Ligand/FLT3L ELISA kit is to be used to detect and quantify protein levels of endogenous mouse or rat Flt-3 Ligand/FLT3L.

This kit contains a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.

  • Antibody specific for mouse or rat Flt-3 Ligand/FLT3L has been pre-coated onto the microwells
  • Mouse or rat Flt-3 Ligand/FLT3L protein in samples is captured by the coated antibody after incubation
  • Following extensive washing, another antibody of biotinylated specific for mouse or rat Flt-3 Ligand/FLT3L is added to detect the captured mouse or rat Flt-3 Ligand/FLT3L protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent

Specifications

Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 3 pg/mL
Assay Sandwich ELISA
Immunoassay Kit Format 96-well Plate
Sample Type Serum
Host Species Mouse, Rat
Quantity 1 Kit
compliance-icons compliance-icons
Product Identifier
  • 96TEST Mouse/Rat Flt-3 Ligand/FLT3L ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.