missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat FABP1/L-FABP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1565-SP
This item is not returnable.
View return policy
Description
FABP1/L-FABP Monoclonal specifically detects FABP1/L-FABP in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| FABP1/L-FABP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P02692 | |
| FABP1 | |
| E. coli-derived recombinant rat FABP1/L-FABP Met1-Ile127 Accession # P02692 | |
| 25 μg | |
| Primary | |
| Detects rat FABP1/L-FABP in direct ELISAs and Western blots. In direct ELISAs and Western blots no cross-reactivity with recombinant rat FABP2, recombinant human FABP3, recombinant mouse (rm) FABP4, or rmFABP5 is observed. | |
| Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 220119 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, LFABP, L-FABP, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2168 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction