missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat DLL1 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF5026
This item is not returnable.
View return policy
Description
DLL1 Polyclonal specifically detects DLL1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| DLL1 | |
| Polyclonal | |
| Unconjugated | |
| Q61483 | |
| DLL1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse DLL1 Ser22-Gln516 Accession # Q61483 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat DLL1 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) DLL1 and less than 1% cross-reactivity with rhDLL3, rhDLL4, and recombinant mouse DLL4 is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Delta, delta (Drosophila)-like 1, Delta 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 28514 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction