missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat DDT Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8354-SP
This item is not returnable.
View return policy
Description
DDT Polyclonal specifically detects DDT in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| DDT | |
| Polyclonal | |
| Western Blot 1 ug/mL | |
| O35215 | |
| DDT | |
| E. coli-derived recombinant mouse DDT Pro2-Leu118 Accession # O35215 | |
| 25 μg | |
| Primary | |
| Detects mouse and rat DDT in direct ELISAs and Western blots. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DDCT, D-dopachrome decarboxylase, D-dopachrome tautomeraseDDCT, DOPD, EC 4.1.1.84, MIF2, Phenylpyruvate Tautomerase II | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1652 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction