missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat DARC Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6695
This item is not returnable.
View return policy
Description
DARC Polyclonal specifically detects DARC in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| DARC | |
| Polyclonal | |
| Unconjugated | |
| NP_034175 | |
| DARC | |
| E. coli-derived recombinant mouse DARC Met1-Pro61, Ala115-Cys127, Ser186-Lys205, Tyr264-Asn285 Accession # NP_034175 | |
| 100 μg | |
| Primary | |
| Detects mouse and rat DARC in Western blots. | |
| Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
| ACKR1, CCBP1, CD234, CD234 antigen, Dfy, Duffy Antigen, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy Antigen, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPD, GPDWBCQ1, GpFy, Plasmodium vivax receptor | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2532 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction