missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse/Rat Cystatin C Quantikine ELISA Kit

Product Code. 18702154
Change view
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18702154 1 x 96-well Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702154 Supplier R&D Systems™ Supplier No. MSCTC0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse/Rat Cystatin C Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat Cystatin C Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat Cystatin C in cell culture supernates (50 ul), cell lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 125.00 - 8000pg/mL
Assay Sensitivity 12.9pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Mouse and Rat Cystatin C
For Use With (Application) ELISA
Gene ID (Entrez) 13010
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ARMD11, bA218C14.4 (cystatin C), CST3, cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, Post-gamma-globulin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.