missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat Complement Component C3d Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 18 publications
Brand: R&D Systems AF2655
This item is not returnable.
View return policy
Description
Complement Component C3d Polyclonal specifically detects Complement Component C3d in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Complement Component C3d | |
| Polyclonal | |
| Unconjugated | |
| P01027 | |
| E. coli-derived recombinant mouse Complement Component C3d His1002-Arg1303 (Cys1010Ser) Accession # P01027 | |
| 100 μg | |
| Primary | |
| Detects mouse Complement Component C3d in direct ELISAs and Western blots. Detects mouse and rat Complement Component C3d in immunohistochemistry. | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 718 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction