missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat CD47 N-terminal IgV-like Extracellular Domain Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF1866-SP
This item is not returnable.
View return policy
Description
CD47 Polyclonal specifically detects CD47 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CD47 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAP, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 961 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.25-1 ug/mL, Immunohistochemistry 1-15 ug/mL | |
| NP_034711 | |
| CD47 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD47 N-terminal IgV-like Gln19-Pro158 Accession # NP_034711 | |
| 25 μg | |
| Primary | |
| Detects mouse CD47 N-terminal IgV-like in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human CD47 is observed. | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction