missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse/Rat CCL5/RANTES Quantikine ELISA Kit
Description
The Mouse/Rat CCL5/RANTES Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat CCL5/RANTES in cell culture supernates (50 ul), serum (25 ul), platelet-poor edta plasma (25 ul), platelet-poor citrate plasma (25 ul).
Specifications
Specifications
| Assay Range | 7.80 - 500pg/mL |
| Assay Sensitivity | 2pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Platelet-poor Plasma (EDTA, Citrate), Serum |
| Specificity | Natural and Recombinant Mouse and Rat RANTES. This assay cross-reacts 1.3% with Recombinant Human RANTES. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 20304 |
| Quantity | 2 x 96-well Plates |
| Sample Volume | Cell Culture Supernates (50uL),Serum (25uL),Platelet-poor EDTA Plasma (25uL),Platelet-poor Citrate Plasma (25uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?