missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse RAGE Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB11795-SP
This item is not returnable.
View return policy
Description
AGER Monoclonal specifically detects AGER in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| RAGE | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| NP_031451 | |
| Ager | |
| Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Leu342 Accession # NP_031451 | |
| 25 μg | |
| Primary | |
| Detects mouse RAGE in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant canine RAGE and no cross-reactivity with recombinant human RAGE or recombinant rat RAGE is observed. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 697023 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5891 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction