missing translation for 'onlineSavingsMsg'
Learn More

Mouse Rae-1 Pan Specific Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 18 publications

£167.00 - £1765.00

Specifications

Antigen Rae-1
Clone 186107
Concentration LYOPH
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Applications Flow Cytometry, CyTOF
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18793939
GREENER_CHOICE
View Documents
R&D Systems
MAB17582-SP
25 μg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18743319
GREENER_CHOICE
View Documents
R&D Systems
MAB17582-100
100 μg
£446.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18714938
GREENER_CHOICE
View Documents
R&D Systems
MAB17582-500
500 μg
£1765.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rae-1 Monoclonal specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

Rae-1
LYOPH
Flow Cytometry, CyTOF
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
8480
Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 Accession # Q9JI58
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Rae-1. It recognizes Rae-1 alpha , beta , delta , gamma and epsilon .
186107
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Monoclonal
Rat
Mouse
Q9JI58
Raet1a
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.