missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse R-Spondin 1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF3474
This item is not returnable.
View return policy
Description
R-Spondin 1 Polyclonal specifically detects R-Spondin 1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| R-Spondin 1 | |
| Polyclonal | |
| Unconjugated | |
| Q9Z132 | |
| RSPO1 | |
| E. coli-derived recombinant mouse R-Spondin 1 (R&D Systems, Catalog # 3474-RS) Ser21-Gly209 Accession # Q9Z132 | |
| 100 μg | |
| Primary | |
| Detects mouse R-Spondin 1 in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant human R-Spondin 1 is observed and less than 5% cross-reactivity with recombinant mouse (rm) R-Spondin 3 and rmR-Spondin 4 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Cristin 3, CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPO1, RSPONDIN, RSpondin 1, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 284654 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction