missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pure-Blot anti-Rabbit IgG (H+L) Secondary Antibody (eB182), HRP, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG Monoclonal antibody specifically detects IgG in Rabbit samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | IgG |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | eB182 |
| Conjugate | HRP |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.1 mg/ml Bovine Serum Albumin (BSA, IgG and Protease free), 50% (v/v) Glycerol |
| Gene Alias | IP Detection Reagent |
| Host Species | Mouse |
| Immunogen | Rabbit IgG |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?