missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pure-Blot anti-Rabbit IgG (H+L) Secondary Antibody (eB182), FITC, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Monoclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Fluorophore-linked immunosorbent assay.
Specifications
Specifications
| Antigen | IgG |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ELISA |
| Classification | Monoclonal |
| Clone | eB182 |
| Concentration | LYOPH |
| Conjugate | FITC |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:2000 - 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 - 1:2500, Immunoprecipitation, Fluorophore-linked immunosorbent assay |
| Formulation | Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/ml Polyethylene Glycol (PEG-8000) |
| Gene Alias | IP Detection Reagent |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?