missing translation for 'onlineSavingsMsg'
Learn More

Mouse Pure-Blot anti-Rabbit IgG (H+L) Secondary Antibody (eB182), DyLight 800, Novus Biologicals™

Product Code. 18389971 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389971 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389971 Supplier Novus Biologicals Supplier No. NBP311666

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG Monoclonal antibody specifically detects IgG in Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ELISA
Classification Monoclonal
Clone eB182
Concentration LYOPH
Conjugate DyLight 800
Dilution Western Blot 1:1000, Flow Cytometry 1:2000 - 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 - 1:2500, Immunoprecipitation, Fluorophore-linked immunosorbent assay
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/ml Polyethylene Glycol (PEG-8000)
Gene Alias IP Detection Reagent
Host Species Mouse
Immunogen Rabbit IgG
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Reconstitution Reconstitute with 100 ul deionized water (or equivalent).
Target Species Rabbit
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.