missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pure-Blot anti-Goat IgG (H+L) Secondary Antibody (eB270), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11670
This item is not returnable.
View return policy
Description
IgG Monoclonal antibody specifically detects IgG in Goat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| IgG | |
| Monoclonal | |
| HRP | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.1 mg/ml Bovine Serum Albumin (BSA, IgG and Protease free), 50% (v/v) Glycerol | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Secondary | |
| Goat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| eB270 | |
| Western Blot 1:000, Immunoprecipitation | |
| IP Detection Reagent | |
| Goat IgG | |
| 200 μg | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| 3500 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction