missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PU.1/Spi-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7124-SP
This item is not returnable.
View return policy
Description
PU.1/Spi-1 Monoclonal specifically detects PU.1/Spi-1 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| PU.1/Spi-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P17433 | |
| Spi1 | |
| E. coli-derived recombinant mouse Spi-1 Met1-Lys169 Accession # P17433 | |
| 25 μg | |
| Primary | |
| Detects mouse PU.1/Spi-1 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human PU.1/Spi-1 is observed. Detects mouse and rat PU.1/Spi-1 in Western Blots | |
| Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 823123 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hematopoietic transcription factor PU.1, OF, PU.1, SFPI1, SFPI131 kDa transforming protein, SPI1, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6688 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering