missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pro EGF Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5256-SP
This item is not returnable.
View return policy
Description
EGF Monoclonal specifically detects EGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| EGF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P01132 | |
| EGF | |
| Mouse myeloma cell line NS0-derived recombinant mouse EGF Trp29-Arg1029 Accession # P01132 | |
| 25 μg | |
| Primary | |
| Detects mouse Pro EGF in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Amphiregulin, rmEpigen, rmEpiregulin, rmEGF, rmMFG-E8, recombinant human (rh) EGF, rhHB-EGF, rhHRG alpha, rhHRG beta, or recombinant rat EGF is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 463004 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1950 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction