missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PRAT4A Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4429-SP
This item is not returnable.
View return policy
Description
PRAT4A/TNRC5 Polyclonal specifically detects PRAT4A/TNRC5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PRAT4A/TNRC5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAG4A, canopy 3 homolog (zebrafish), CNPY3, CTG4A, ERDA5, ERDA5CTG repeat protein 4a, Expanded repeat-domain protein CAG/CTG 5, PRAT4ACAG repeat containing, Protein associated with TLR4, protein associated with Toll-like receptor 4A, protein canopy homolog 3, TNRC5, TNRC5expanded repeat domain, CAG/CTG 5, trinucleotide repeat containing 5, Trinucleotide repeat-containing gene 5 protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10695 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9DAU1 | |
| CNPY3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PRAT4A isoform 1 Glu38-Leu276 Accession # Q9DAU1 | |
| 25 μg | |
| Primary | |
| Detects mouse PRAT4A in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse PRAT4B is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction