missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Podocan Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3104-SP
This item is not returnable.
View return policy
Description
Podocan Polyclonal specifically detects Podocan in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Podocan | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PCAN, PODN, PODOCAN, podocan proteoglycan, SLRR5A, SLRR5AMGC24995 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 127435 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q7TQ62 | |
| PODN | |
| Mouse myeloma cell line NS0-derived recombinant mouse Podocan Val24-Arg611 Accession # Q7TQ62 | |
| 25 μg | |
| Primary | |
| Detects mouse Podocan in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Podocan is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction