Learn More
R&D Systems™ Mouse Plexin B3 (NP_062533.2) VersaClone cDNA
Product Code. 18703047
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity Unit Size
18703047 10 μg 10µg
1 options
Product Code. 18703047 Supplier R&D Systems™ Supplier No. RDC3697
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Gene Plexin B3
Accession Number NP_062533.2
Gene ID (Entrez) 5365
Gene Alias PLEXB3, plexin B3, plexin-B3, PLEXR
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Line R&D Systems