missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PIR-B Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB2754
This item is not returnable.
View return policy
Description
PIR-B Monoclonal specifically detects PIR-B in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| PIR-B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| AAH26937 | |
| Pirb | |
| Mouse myeloma cell line NS0-derived recombinant mouse PIR-B Gly24-Gly635 Accession # AAH26937 | |
| 100 μg | |
| Primary | |
| Detects mouse PIR-B in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse PIR-A. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 326414 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD85a, HL9, ILT5, ILT-5, leukocyte immunoglobulin like receptor B3, LILRA6, LILRB3, LIR3, LIR-3, PIRB, PIR-B | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11025 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction