missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PIR-B Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB2754S-100UG
This item is not returnable.
View return policy
Description
PIR-B Monoclonal specifically detects PIR-B in Mouse samples. It is validated for Flow Cytometry.
Specifications
| PIR-B | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAH26937 | |
| Pirb | |
| Mouse myeloma cell line NS0-derived recombinant mouse PIR-B Gly24-Gly635 Accession # AAH26937 | |
| RUO | |
| 11025 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 326414 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD85a, HL9, ILT5, ILT-5, leukocyte immunoglobulin like receptor B3, LILRA6, LILRB3, LIR3, LIR-3, PIRB, PIR-B | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse PIR-B in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse PIR-A. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction