missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse Pentraxin 3/TSG-14 ELISA Kit

Product Code. 17761499 Shop All Proteintech Products
Change view
Click to view available options
Immunoassay Kit Format:
96-well Plate
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Immunoassay Kit Format unitSize
17761499 96-well Plate 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17761499 Supplier Proteintech™ Supplier No. KE1007396T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Mouse Pentraxin 3/TSG-14 ELISA kit is to be used to detect and quantify protein levels of endogenous mouse Pentraxin 3/TSG-14.

This kit contains a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.

  • Antibody specific for mouse Pentraxin 3/TSG-14 has been pre-coated onto the microwells
  • Mouse Pentraxin 3/TSG-14 protein in samples is captured by the coated antibody after incubation
  • Following extensive washing, another antibody of biotinylated specific for mouse Pentraxin 3/TSG-14 is added to detect the captured mouse Pentraxin 3/TSG-14 protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent

Specifications

Assay Sandwich ELISA
Immunoassay Kit Format 96-well Plate
Sample Type Cell Culture Supernatant, Serum
Gene ID (Entrez) 19288
Host Species Mouse
compliance-icons compliance-icons
Product Identifier
  • 96TEST Mouse Pentraxin 3/TSG-14 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.