missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PEDFR/PNPLA2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5365-SP
This item is not returnable.
View return policy
Description
PEDFR/PNPLA2/ATGL Monoclonal specifically detects PEDFR/PNPLA2/ATGL in Mouse samples. It is validated for Western Blot.
Specifications
| PEDFR/PNPLA2/ATGL | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q8BJ56 | |
| PNPLA2 | |
| E. coli-derived recombinant mouse PEDF R/PNPLA2 Val162-Gly253 Accession # Q8BJ56 | |
| 25 μg | |
| Primary | |
| Detects PEDF R/PNPLA2 in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 615728 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adipose triglyceride lipase, ATGL, ATGL1110001C14Rik, Calcium-independent phospholipase A2, Desnutrin, EC 3.1.1.3, FP17548, IPLA2-zeta, patatin-like phospholipase domain containing 2, patatin-like phospholipase domain-containing protein 2, patatin-like phospholipase domain-containing protein 2-like, PEDF R, PEDF-R, Pigment epithelium-derived factor, PNPLA2, Transport-secretion protein 2, transport-secretion protein 2.2, triglyceride hydrolase, TTS2, TTS2.2, TTS-2.2, TTS2DKFZp667M109 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57104 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction