missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PDGF R beta Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB1042
This item is not returnable.
View return policy
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Western Blot.
Specifications
| PDGF R beta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P05622 | |
| PDGFRB | |
| Mouse myeloma cell line NS0-derived recombinant mouse PDGF R beta Leu32-Lys530 Accession # P05622 | |
| 500 μg | |
| Primary | |
| Detects mouse PDGF R beta in direct ELISAs and Western blots. In direct ELISAs, approximately 10-20% cross-reactivity with recombinant human PDGF R beta is observed, but no cross-reactivity with recombinant mouse (rm) PDGF R alpha , rmFlt-3, rmSCF R, or rmLymphotoxin beta R is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 182106 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5159 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction