missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PDGF R alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1062-SP
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Mouse samples. It is validated for Western Blot.
Specifications
| PDGFR alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P26618 | |
| PDGFRA | |
| Mouse myeloma cell line NS0-derived recombinant mouse PDGF R alpha Leu25-Glu524 (Asp65Glu, Gly439Ala, Thr440Ala), predicted Accession # P26618 | |
| 25 μg | |
| Primary | |
| Detects mouse PDGF R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, 5-20% cross-reactivity with recombinant human PDGF R alpha is observed and no cross-reactivity with recombinant mouse PDGF R beta is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 189208 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5156 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction