missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PD-L1/B7-H1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1019-SP
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal specifically detects PD-L1 in Mouse samples. It is validated for Western Blot.
Specifications
| PD-L1/B7-H1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9EP73 | |
| CD274 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PD-L1/B7-H1 Phe19-Thr238 Accession # Q9EP73 | |
| 25 μg | |
| Primary | |
| Detects mouse PD-L1/B7-H1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) B7-1, rmB7-2, recombinant human (rh) B7-H1, rmB7-H2, rmB7-H3, rmB7-H4, or rmPD-L2 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 179711 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 29126 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction