missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Patched 1/PTCH (First Extracellular Loop) Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4105-SP
This item is not returnable.
View return policy
Description
Patched 1/PTCH Polyclonal specifically detects Patched 1/PTCH in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Patched 1/PTCH | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BCNS, BCNSFLJ42602, FLJ26746, HPE7, mes, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4', PTCH protein -10, PTCH1, PTCH11 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5727 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q61115 | |
| PTCH1 | |
| E. coli-derived recombinant mouse Patched 1/PTCH Glu108-Asp422 Accession # Q61115 | |
| 25 μg | |
| Primary | |
| Detects mouse Patched 1/PTCH in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) Patched 1 is observed, and less than 1% cross-reactivity with rhPatched-2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction