Learn More
Mouse p53 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB1746
Product Code. 18159710
  • £399.00 / 100µg
Description
p53 Monoclonal specifically detects p53 in Mouse samples. It is validated for Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p53
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
7157
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b
Immunoprecipitation
197643
Immunoprecipitation 1-3 ug/500 ug cell lysate
NP_035770
Trp53
E. coli-derived recombinant mouse p53 Leu28-Phe325 Accession # NP_035770
100 μg
Primary
Detects mouse p53 in Western blots.
Mouse
Purified