missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse P-Selectin/CD62P Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF737
This item is not returnable.
View return policy
Description
P-Selectin/CD62P Polyclonal specifically detects P-Selectin/CD62P in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| P-Selectin/CD62P | |
| Polyclonal | |
| Biotin | |
| Q01102 | |
| SELP | |
| Mouse myeloma cell line NS0-derived recombinant mouse P-Selectin/CD62P Trp42-Ala709 Accession # Q01102 | |
| 50 μg | |
| Primary | |
| Detects mouse P-Selectin/CD62P in ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human P-Selectin, recombinant mouse (rm) E-Selectin, and rmL-Selectin is observed. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6403 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction