missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OX40 Ligand/TNFSF4 Antibody, R&D Systems™

Rat Monoclonal Antibody
£167.00 - £487.00
Specifications
| Antigen | OX40 Ligand/TNFSF4 |
|---|---|
| Clone | 182609 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18757169
![]() |
R&D Systems
MAB12362-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18130541
|
R&D Systems
MAB12362 |
500 μg |
£487.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| OX40 Ligand/TNFSF4 | |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P43488 | |
| 7292 | |
| Mouse myeloma cell line NS0-derived recombinant mouse OX40 Ligand/TNFSF4 Gln49-Leu198 Accession # P43488 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse OX40 Ligand/TNFSF4 in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) OX40, recombinant human (rh) OX40 Ligand, rmFas Ligand, rmLIGHT, or rhLIGHT is observed. In Western blots, 100% cross-reactivity with rmTNF-alpha is observed, 15-30% with rmTRANCE and rmTRAIL is observed, and no cross-reactivity with rmBAFF, rmEDA, rmGITR Ligand, rmLIGHT, rmTWEAK, rmFas Ligand, rhAPRIL, or rhOX40 Ligand is observed. |
| 182609 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| TNFSF4 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title