missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OX40 Ligand/TNFSF4 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1236-100
This item is not returnable.
View return policy
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| OX40 Ligand/TNFSF4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.01-0.05 ug/mL | |
| P43488 | |
| TNFSF4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse OX40 Ligand/TNFSF4 Gln49-Leu198 Accession # P43488 | |
| 100 μg | |
| Primary | |
| Detects mouse OX40 Ligand/TNFSF4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) APRIL, rhGITR Ligand, rhLIGHT, rhLT alpha 1/beta 2, rhLT alpha 2/beta 1, recombinant cotton rat TNF-alpha , recombinant mouse (rm) TNF-alpha , rmTRAIL, rhTRANCE, rmTRANCE, rmTWEAK, or rhVEGI is observed. | |
| Mouse | |
| Supernatant |
| Western Blot, Neutralization | |
| 182601 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7292 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction