missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Osteopontin/OPN Antibody, R&D Systems™

Rabbit Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB808-100
This item is not returnable.
View return policy
Description
Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Osteopontin/OPN | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
| Mouse myeloma cell line NS0-derived recombinant mouse Osteopontin/OPN Leu17-Asn294 Accession # Q547B5 | |
| 100 μg | |
| 6696 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 2139B | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Q547B5 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects mouse OsteopontinOPN in direct ELISAs. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction