missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Osteoadherin/OSAD Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3308-SP
This item is not returnable.
View return policy
Description
Osteoadherin/OSAD/OMD Monoclonal specifically detects Osteoadherin/OSAD/OMD in Mouse samples. It is validated for Western Blot.
Specifications
| Osteoadherin/OSAD/OMD | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O35103 | |
| OMD | |
| Mouse myeloma cell line NS0-derived recombinant mouse Osteoadherin/OSAD Gln21-Ile423 Accession # O35103 | |
| 25 μg | |
| Primary | |
| Detects mouse Osteoadherin/OSAD in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human OSAD is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 348423 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Keratan sulfate proteoglycan osteomodulin, KSPG osteomodulin, OMD, OSAD, Osteoadherin, osteoadherin proteoglycan, osteomodulin, SLRR2C, SLRR2Costeoadherin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4958 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction